Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF1B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-556 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-556 Supplier Abnova Corporation Supplier No. H00023095A01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse polyclonal antibody raised against a full-length recombinant KIF1B.

This gene encodes a motor protein that transports mitochondria and synaptic vesicle precursors. Mutations in this gene cause Charcot-Marie-Tooth disease, type 2A1. [provided by RefSeq

Sequence: MSKTSLGQSMSKYDLLVWFEISELEPTGEYIPAVVDHTAGLPCQGTFLLHQGIQRRITVTIIHEKGSELHWKDVRELVVGRIRNKPEVDEAAVDAILSLNIISAKYLKSSHNSSRTFYRFEAVWDSSLHNSLLLNRVTPYGEKIYMTLSAYLELDHCIQPAVITKDVCMVFYSRDAKISPPRSLRSLFGSGYSKSPDSNRVTGIYELSLCKMSDTGSPGMQRRRRKILDTSVAYVRGEENLAGWRPRGDSLILEHQWELEKLELLHEVEKTRHFLLLRERLGDSIPKSLSDSLSPSLSSGTLSTSTSISSQISTTTFESAITPSESSGYDSGDIESLVDREKELATKCLQLLTHTFNREFSQVHGSVSDCKLSDISPIGRDPSESSFSSATLTPSSTCPSLVDSRSNSLDQKTPEANSRASSPCPEFEQFQIVPAVETPYLARAGKNEFLNLVPDIEEIRPSSVVSKKGYLHFKEPLYSNWAKHFVVVRRPYVFIYNSDKDPVERGIINLSTAQVEYSEDQQAMVKTPNTFAVCTKHRGVLLQALNDKDMNDWLYAFNPLLAGTIRSKLSRRCPSQSKY

Specifications

Antigen KIF1B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant KIF1B.
Formulation 50% glycerol
Gene KIF1B
Gene Accession No. BC001415
Gene Alias CMT2/CMT2A/CMT2A1/FLJ23699/HMSNII/KIAA0591/KIAA1448/KLP/MGC134844
Gene Symbols KIF1B
Host Species Mouse
Immunogen KIF1B (AAH01415, 1 a.a. ∽ 579 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23095
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.