Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MACF1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106613 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-613 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-613 Supplier Abnova Corporation Supplier No. H00023499A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MACF1.

The protein encoded by this gene belongs to the plakin family of cytoskeletal linker proteins. This protein family forms bridges between different cytoskeletal elements through specialized modular domains. The encoded protein is one of the largest size proteins identified in human cytoskeletal proteins. It has functional actin and microtubule binding domains, and it appears to stabilize actin at sites where microtubules and microfilaments meet. It may function in microtubule dynamics to facilitate actin-microtubule interactions at the cell periphery and to couple the microtubule network to cellular junctions. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq

Sequence: KIEDEVTRQVAQCKCAKRFQVEQIGENKYRFGDSQQLRLVRILRSTVMVRVGGGWMALDEFLVKNDPCRARGRTNIELREKFILPEGASQGMTPF

Specifications

Antigen MACF1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MACF1.
Formulation 50% glycerol
Gene MACF1
Gene Accession No. BC007330
Gene Alias ABP620/ACF7/FLJ45612/FLJ46776/KIAA0465/KIAA1251/MACF/OFC4
Gene Symbols MACF1
Host Species Mouse
Immunogen MACF1 (AAH07330, 1 a.a. ∽ 95 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23499
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.