Learn More
Cell Sciences Inc Mouse Anti-Monkey CD3 (Clone FN-18) mAb, 500ug
Supplier:  Cell Sciences Inc CMM500
Encompass_Preferred
Mouse Anti-Monkey CD3 Clone FN-18 monoclonal antibody is able to stimulate rhesus macaque PBMC. This clone cross reacts with cynomolgus monkey and baboon CD3.
Catalog No. NC2652306
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.