Learn More
Invitrogen™ Mouse anti-Monkey IgG (H+L) Secondary Antibody
Mouse Monoclonal Secondary Antibody
Supplier:  Invitrogen™ SA110653
Catalog No. 01-671-620
Only null left
Add to Cart
Description
Description
SA1-10653 detects monkey IgG, and will not cross-react with human, bovine, equine or ovine IgG. SA1-10653 has been successfully used in ELISA applications. The SA1-10653 immunogen is purified Rhesus monkey IgG.
Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkey IgG (H+L)
Monoclonal
1.3 mg/mL
PBS with no preservative; pH 7.4
Purified Rhesus monkey IgG.
500 μg
Secondary
-20°C
Liquid
ELISA
5C12.D4
Unconjugated
Mouse
Protein G
RUO
Non-human Primate
Secondary Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.