Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64 a and b Alloantigens Mouse anti-Mouse, Brilliant Violet 650, Clone: X54-5/7.1, BD Optibuild™

Catalog No. BDB740622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740622 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740622 Supplier BD Biosciences Supplier No. 740622
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The X54-5/7.1 monoclonal antibody specifically recognizes FcγRI (CD64) encoded by the more common Fcgr1a and Fcgr1b alleles. The alloantigens generated by the Fcgr1a and Fcgr1b alleles, have been confirmed positive in mouse strains BALB/c and C57BL/6 and reported positive in strains 129, A, AKR, ALR, BUB, C3H, C57BL/10, C57BLKS, C57BR, C58, CBA, CE, DBA/2, HRS, MRL, NON, NZB, NZO, NZW, PL, SJL, ST, SWR. The a and b alloantigens have been reported negative in mouse strains ABH, NOD. CD64, a key receptor in the development of immune responses, has a dual role as a low affinity receptor for IgG3 and high affinity receptor for IgG2a linking innate and adaptive immunities. CD64 mediates endocytosis, phagocytosis, antibody-dependent cellular toxicity, cytokine release and superoxide generation. CD64 is expressed largely on macrophages and dendritic cells. For more information regarding clone X54-5/7.1 and the alloantigens it recognizes, please refer to the reference by Tan et al.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD64 a and b Alloantigens
Applications Flow Cytometry
Classification Monoclonal
Clone X54-5/7.1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description Fcgr1; FcRI; Fcg1; Fc-gamma RI; Fc receptor IgG high affinity I; IGGHAFC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse CD64 a Alloantigen
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.