Learn More
Mouse anti-Mouse IgD[a], Clone: AMS 9.1, biotin, Secondary Antibody, BD
Shop All BD Biosciences ProductsDescription
The AMS 9.1 antibody reacts specifically with the Fc portion of mouse IgD of Igh C[a] haplotype (eg, AKR, BALB/c, CBA, C3H/He, C58, DBA/1, DBA/2, NZB). It does not react with IgD of Igh-C[b] or Igh-C[e] haplotypes (eg, C57BL/6, C57BL/10, SJL, A). AMS 9.1
Host Species: Mouse
Clone: AMS 9.1
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: BALB/c mouse splenocytes
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgD[a] |
| Classification | Monoclonal |
| Clone | AMS 9.1 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.