Learn More
Mouse anti-Mouse IgM[b], Clone: AF6-78, PE, Secondary Antibody, BD
Supplier: BD Biosciences 553521
Description
The AF6-78 antibody reacts specifically with mouse IgM of Igh-C[b] and related haplotypes (e.g., C57BL/6, C57BL/10, SJL, AKR, NZB). It does not react with IgM of Igh-C[a] or related haplotypes (e.g., BALB/c, C58, CBA, C3H/Bi, C3H/He, DBA/1, DBA/2, PL, RII
Host Species: Mouse
Clone: AF6-78
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: C57BL/10 mouse splenocytes
ELISA, Flow Cytometry

Specifications
Mouse anti-Mouse IgM[b] | |
AF6-78 | |
PE | |
0.2 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.2mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.