Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-108 Mouse anti-Mouse, Brilliant Violet 421, Clone: 13G3, BD Optibuild™

Catalog No. BDB740090 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740090 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740090 Supplier BD Biosciences Supplier No. 740090
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 13G3 monoclonal antibody specifically binds to Lymphocyte antigen 108, Ly-108. Ly-108 is a member of the signaling lymphocytic activation molecule (SLAM) family of immune receptors. Ly-108 is a type 1 transmembrane glycoprotein adhesion receptor that is expressed by T cells, NKT cells, B cells, NK cells, macrophages, dendritic cells and neutrophils. Ly-108 plays multiple roles in innate and adaptive immunity including costimulation of NK cell cytotoxicity and T cell cytokine responses. Moreover, Ly-108 has been implicated in autoimmunity.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-108
Applications Flow Cytometry
Classification Monoclonal
Clone 13G3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Slamf6; SLAM family member 6; KAL1; NTB-A; SF2000
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen WT thymocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.