Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-N615, Clone: P5E2, Abnova™

Catalog No. 89035595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-595 Supplier Abnova Corporation Supplier No. MAB14096

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human IgM.

  • High specificity
  • High reactivity

Specifications

Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone P5E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human IgM.
Dilution ELISA (1 ug/mL) Flow Cytometry (1 ug/mL) Immunoprecipitation Radioimmunoassay Western Blot (1:100-1:400) The optimal working dilution should be determined by the end user.
Formulation Liquid
Host Species Mouse
Immunogen Purified Human Serum IgM.
Purification Method Ion exchange chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.