Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Ninjurin-1/NINJ1, Clone: 12H6.1,

Catalog No. MABN816 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN816 Supplier MilliporeSigma™ Supplier No. MABN816
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Ninjurin-1/NINJ1 clone: 12H6.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Ninjurin-1 (UniProt Q92982; also known as Nerve injury-induced protein 1) is encoded by the NINJ1 (also known as NIN1, NINJURIN) gene (Gene ID 4814) in human. Ninjurin-1 (NINJ1) is a cell surface adhesion molecule originally identified to be up-regulated in neuronal and Schwann cells after sciatic nerve injury. NINJ1 is also reported to play a role in the progression of multiple sclerosis, an autoimmune inflammatory CNS disease characterized by demyelination and axonal damage. In addition, NINJ1 is known to cause regression of hyaloid blood vessels important for the maturation of the lens during embryonic development. Consistent with the latter role in vascular system, Penile injection of Ninj1-neutralizing antibody is shown to significantly restore erectile function in a murine model of diabetic erectile disfunction (ED) as a result of enhanced cavernous endothelial cell proliferation. Human Ninjurin-1 is a 152-amino aicd protein with boht its N- and C-terminal ends exposed extracellulary (a.a. 1-80, 142-152) and a short intracellular segment (a.a. 102-119) sandwiched between two transmembrane domains (a.a. 81-101, 121-141).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninjurin-1/NINJ1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 12H6.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q92982
Gene Symbols NINJ1; NIN1; NINJURIN
Host Species Mouse
Immunogen GST-tagged recombinant human Ninjurin-1/NINJ1 N-terminal extracellular domain.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_004139
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.