Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PERK, Clone: 4F6.1,

Catalog No. MABN842MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN842MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN842MI Supplier MilliporeSigma™ Supplier No. MABN842
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PERK clone: 4F6.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Eukaryotic translation initiation factor 2-alpha kinase 3 (EC 2.7.11.1; UniProt Q9NZJ5; also known as Eukaryotic translation initiation factor 2 alpha kinase 3, HsPEK, Pancreatic EIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase) is encoded by the EIF2AK3 (also known as PEK, PERK, WRS) gene (Gene ID 9451) in human. The accumulation of unfolded or misfolded proteins in the endoplasmic reticulum (ER) induces the activation of three main unfold protein response (UPR) pathways, the inositol requiring enzyme-1 (IRE1) pathway, the activating transcription factor 6 (ATF6) pathway, and the protein kinase-like ER kinase (PERK) pathway. The PERK pathway involves PERK auto-phosphorylation/activation, resulting in PERK phosphorylation of eIF2α, which in turn leads to increased ATF4 transcription and ATF4-mediated CAAT/enhancer binding protein (C/EBP) homologous protein (CHOP) upregulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PERK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4F6.1
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9NZJ5
Gene Symbols EIF2AK3; PEK; PERK
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the Internal (C-terminal half) of human PERK.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_004827
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.