Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Diagnostics Mouse Anti-PNU-159682 Monoclonal Antibody, clone G8G3, IgG, Unconjugated, 500 μL

Supplier:  Creative Diagnostics CABTL3111500UL

Encompass

This Mouse Anti-PNU-159682 monoclonal antibody, clone G8G3 from Creative Diagnostics, is designed for high specificity in detecting PNU-159682. It is an unconjugated IgG antibody validated for use in ELISA applications, making it a reliable choice for researchers needing precise target identification.

  • Monoclonal antibody clone: G8G3
  • Isotype: IgG
  • Conjugate: Unconjugated
  • Host species: Mouse
  • Species reactivity: N/A
  • Applications: ELISA
  • Target: PNU-159682
  • Alternative target names: PNU159682, PNU 159682
  • Quantity: 500 μL

Catalog No. NC2163613


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.