Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-PP2A, C subunit, Clone: 1D6, Alexa Fluor 488,

Catalog No. 05421AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05-421-AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-421-AF488 Supplier MilliporeSigma™ Supplier No. 05421AF488
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PP2A, C subunit clone: 1D6 in Bovine, Human, Mouse, Rabbit, Rat, Xenopus, Yeast samples, and it is validated for Immunocytochemistry

Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform (EC 3.1.3.16; UniProt P67775; PP2A-alpha, PP2Ac, PP2Calpha, Replication protein C, RP-C) is encoded by the PPP2CA (also known as PP2CA) gene (Gene ID 5515) in human. Protein phosphatase 2 (PP2 or PP2A) is a ubiquitously expressed heterotrimeric serine/threonine protein phosphatase with broad substrate specificity, such as Raf, MEK, and AKT. PP2A is composed of a structural A and a catalytic C subunit as the dimeric core enzyme, and a regulatory B subunit. There exist two different A subunits (PPP2R1A & PPP2R1B), two different C subunits (PPP2CA & PPP2CB), and 12 different B subunits (PPP2R2B-D, PPP2R3A-C, PPP2R4, PPP2R5A-E). The enzymatic activity of a catalytic subunit is altered upon binding an A subunit even in the absence of a B subunit. The B subunits are believed to play key roles in controlling the localization and specific activity of the A/C holoenzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A, C subunit
Applications Immunocytochemistry
Classification Monoclonal
Clone 1D6
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG2bκ antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P67775
Gene Symbols PPP2CA; PP2CA
Host Species Mouse
Immunogen A 16 residue synthetic peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of the 36kDa catalytic subunit of human protein phosphatase 2A (PP2A).
Purification Method Protein A Purfied
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002706
Target Species Bovine, Human, Mouse, Rabbit, Rat, Xenopus species, Yeast
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.