Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Protocadherin-19 APC-conjugated Antibody, R&D Systems™

Catalog No. FAB8626A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8626A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8626A025 Supplier R&D Systems Supplier No. FAB8626A025
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protocadherin-19 Monoclonal specifically detects Protocadherin-19 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Protocadherin-19
Applications Flow Cytometry
Classification Monoclonal
Clone 921614
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8TAB3
Gene Symbols PCDH19
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Protocadherin-19 Met1-Ser678 Accession # Q8TAB3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57526
Test Specificity Detects human Protocadherin-19 in direct ELISA and flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.