Learn More
Mouse anti-Rat IgA Heavy Chain Secondary Antibody (MARA-1), DyLight 550, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MARA-1 |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified IR1060 IgA rat myeloma protein |
| Purification Method | Affinity purified |
| Quantity | 0.25 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.