Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgA Heavy Chain Secondary Antibody (MARA-1), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB552217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB552217 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB552217 Supplier Novus Biologicals Supplier No. NBP268392DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgA Heavy Chain Monoclonal specifically detects IgA Heavy Chain in Rat samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen IgA Heavy Chain
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MARA-1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified IR1060 IgA rat myeloma protein
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.