Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgD Secondary Antibody (MARD-3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB552945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB552945 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB552945 Supplier Novus Biologicals Supplier No. NBP268388V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen IgD
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MARD-3
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Host Species Mouse
Immunogen IR731 myeloma protein
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3495
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.