Learn More
Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), DyLight 594, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgE Heavy Chain |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | MARE-1 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon |
| Host Species | Mouse |
| Immunogen | Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.