Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), PE, R&D Systems™
SDP

Catalog No. NB554412 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554412 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554412 Supplier R&D Systems Supplier No. NBP268394PE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgE Heavy Chain Monoclonal specifically detects IgE Heavy Chain in Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgE Heavy Chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone MARE-1
Conjugate PE
Formulation PBS
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Host Species Mouse
Immunogen Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.