Learn More
Description
The G28-5 antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. A suspension of pooled rat IgG1 was used as the source of immunogen.
Host Species: Mouse
Clone: G28-5
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Pooled rat IgG
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Rat IgG1/2a |
| Classification | Monoclonal |
| Clone | G28-5 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.