Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgG1 Heavy Chain Secondary Antibody (MARG1-2), R&D Systems™
SDP

Catalog No. NB002831 Shop All Novus Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002831 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002831 Supplier R&D Systems Supplier No. NBP268397250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG1 Heavy Chain Monoclonal specifically detects IgG1 Heavy Chain in Rat samples. It is validated for ELISA.

Specifications

Antigen IgG1 Heavy Chain
Applications ELISA
Classification Monoclonal
Clone MARG1-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 3 μg/mL - 5 μg/mL
Formulation PBS
Host Species Mouse
Immunogen Rat IR27 IgG1 myeloma protein
Purification Method Affinity Purified
Quantity 250 μg
Regulatory Status RUO
Test Specificity Mouse anti rat IgG1, clone MARG1-2, recognizes the heavy chain of the rat immunoglobulin G1 subclass. No cross-reactivity has been found with other rat immunoglobulin classes or subclasses.
Target Species Rat
Content And Storage Store at 4°C.
Product Type Antibody
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.