Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Rat IgG1 Secondary Antibody, PE, eBioscience™

Mouse Monoclonal Secondary Antibody

$243.00

Specifications

Antigen Rat IgG1
Clone R1-12D10
Concentration 0.2 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-9274
Encompass_Preferred
View Documents
Invitrogen™
12481282
100 μg
Each of 1 for $243.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The R1-12D10 monoclonal antibody recognizes rat IgG1 antibodies and can be used as a second step reagent in microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG1. Applications Reported: This R1-12D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R1-12D10 antibody has been tested by flow cytometric analysis of mouse splenocytes stained with a rat IgG1 primary antibody. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rat IgG1
0.2 mg/mL
Flow Cytometry
PE
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
Secondary Antibody
R1-12D10
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG
Rat
Secondary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.