Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgG2a Heavy Chain Secondary Antibody (MARG2a-1), R&D Systems™
SDP

Catalog No. NB002834 Shop All Novus Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002834 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002834 Supplier R&D Systems Supplier No. NBP268423250UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG2a Heavy Chain Monoclonal specifically detects IgG2a Heavy Chain in Rat samples. It is validated for ELISA.

Specifications

Antigen IgG2a Heavy Chain
Applications ELISA
Classification Monoclonal
Clone MARG2a-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Host Species Mouse
Immunogen Purified rat IgG2a
Quantity 250 μg
Regulatory Status RUO
Test Specificity Mouse anti Human beta-2 Adrenergic Receptor antibody, clone R11E1, recognizes human beta 2 adrenergic receptor (beta2AR), a member of the G protein-coupled receptor superfamily.
Target Species Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.