Learn More
Description
The G15-337 antibody reacts specifically with rat IgG2b. It does not react with other Ig isotypes. A suspension of pooled rat Ig was used as the source of immunogen.
Host Species: Mouse
Clone: G15-337
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat Ig
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Mouse anti-Rat IgG2b |
| Classification | Monoclonal |
| Clone | G15-337 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.