Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Rat IgG2b (Heavy chain) Secondary Antibody, FITC

Catalog No. SA125267
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA125267 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA125267 Supplier Invitrogen™ Supplier No. SA125267

Mouse Monoclonal Secondary Antibody

SA1-25267 detects rat IgG2b, Heavy Chain in flow cytometry applications. Store antibody at 4°C short term, and -20°C long term. Antibody is photosensitive; store in the dark.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2b (Heavy chain)
Applications Flow Cytometry
Classification Monoclonal
Clone MARG2b-3
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.09% sodium azide
Host Species Mouse
Immunogen Rat IgG2b.
Purification Method Affinity chromatography
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.