Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rat IgG2b Heavy Chain Secondary Antibody (MARG2b-8), FITC, Novus Biologicals™
SDP

Catalog No. NB554617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554617 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554617 Supplier Novus Biologicals Supplier No. NBP268401F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG2b Heavy Chain Monoclonal specifically detects IgG2b Heavy Chain in Rat samples. It is validated for ELISA.

Specifications

Antigen IgG2b Heavy Chain
Applications ELISA
Classification Monoclonal
Clone MARG2b-8
Conjugate FITC
Formulation PBS
Host Species Mouse
Immunogen IR863 rat IgG2b myeloma protein
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.