Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody
Mouse Monoclonal Secondary Antibody
$413.50 - $471.00
Specifications
Antigen | Rat Kappa Light Chain |
---|---|
Clone | OX-12 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
SA170012
|
Invitrogen™
SA170012 |
250 μg |
Each for $413.50
|
|
|||||
PISA510177
|
Invitrogen™
SA510177 |
500 μg |
Each for $471.00
|
|
|||||
Description
Mouse anti-Rat Kappa Light Chain Secondary Antibody for Flow
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat Kappa Light Chain | |
1 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.02% sodium azide | |
Pooled rat serum IgG. | |
Secondary Antibody |
OX-12 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Rat | |
Iglc | |
Secondary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title