Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF123, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107256 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-256 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-256 Supplier Abnova Corporation Supplier No. H00063891A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant RNF123.

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq

Sequence: ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAA

Specifications

Antigen RNF123
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant RNF123.
Formulation 50% glycerol
Gene RNF123
Gene Accession No. NM_022064
Gene Alias DKFZp686C2222/FLJ12565/FP1477/KPC1/MGC163504
Gene Symbols RNF123
Host Species Mouse
Immunogen RNF123 (NP_071347, 1216 a.a. ∽ 1314 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63891
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.