Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAR1B, Mouse, Clone: AT1C7, Abnova™

Catalog No. 89109376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-376 Supplier Abnova Corporation Supplier No. MAB11207
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SAR1B.

Specifications

Antigen SAR1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT1C7
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SAR1B.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.02% Sodium Azide.
Gene SAR1B
Gene Alias CMRD/GTBPB/SARA2
Gene Symbols SAR1B
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-198 of human SAR1B.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51128
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.