Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-428 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-428 Supplier Abnova Corporation Supplier No. H00006427B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SFRS2 protein.

Sequence: MSYGRPPPDVEGMTSLKVDNLTYRTSPDTLRRVFEKYGRVGDVYIPRDRYTKESRGFAFVRFHDKRDAEDAMDAMDGADPGVGAVPGLAADLATAARSLGPALVLDLGRPPSPDPHEGPSPSPRRSPDLVRGPGPGLGPGVLPQCPRGNPNPGRDRRVPPSLLKRKERCPLKKMLRSPV

Specifications

Antigen SFRS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SFRS2 protein.
Formulation PBS with no preservative; pH 7.4
Gene SFRS2
Gene Accession No. BC066958
Gene Alias PR264/SC-35/SC35/SFRS2A/SRp30b
Gene Symbols SFRS2
Host Species Mouse
Immunogen SFRS2 (AAH66958,1 a.a. ∽ 179 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6427
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.