Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A10, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107177 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-177 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-177 Supplier Abnova Corporation Supplier No. H00057181A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SLC39A10.

Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A10 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003 [PubMed 12659941]).[supplied by OMIM

Sequence: CIRMFKHYKQQRGKQKWFMKQNTEESTIGRKLSDHKLNNTPDSDWLQLKPLAGTDDSVVSEDRLNETELTDLEGQQESPPKNYLCIEEEKIIDHSHSDGLHTIHEHDL

Specifications

Antigen SLC39A10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC39A10.
Formulation 50% glycerol
Gene SLC39A10
Gene Accession No. NM_020342
Gene Alias DKFZp781L10106/LZT-Hs2/MGC126565/MGC138428
Gene Symbols SLC39A10
Host Species Mouse
Immunogen SLC39A10 (NP_065075, 514 a.a. ∽ 621 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57181
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.