Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRA1, Mouse, Clone: 1D4H8, Abnova™

Catalog No. 89108595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-595 Supplier Abnova Corporation Supplier No. MAB10409
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant SRA1.

This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a noncoding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer. [provided by RefSeq

Specifications

Antigen SRA1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1D4H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant SRA1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene SRA1
Gene Alias MGC87674/SRA/SRAP/STRAA1/pp7684
Gene Symbols SRA1
Host Species Mouse
Immunogen Recombinant protein corresponding to human SRA1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10011
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.