Learn More
United States Biological Corporation Mouse Anti-Streptavidin, Monoclonal, Mouse, IgG1, 1mg
Supplier:  United States Biological Corporation S797370
Encompass_Preferred
Suitable for use in Immunohistochemistry, Flow Cytometry, ELISA, Immunocytochemistry and Western Blot. Other applications have not been tested.
Catalog No. NC3735435
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.