Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SVIL, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105540 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-540 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-540 Supplier Abnova Corporation Supplier No. H00006840A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SVIL.

This gene encodes a bipartite protein with distinct amino- and carboxy-terminal domains. The amino-terminus contains nuclear localization signals and the carboxy-terminus contains numerous consecutive sequences with extensive similarity to proteins in the gelsolin family of actin-binding proteins, which cap, nucleate, and/or sever actin filaments. The gene product is tightly associated with both actin filaments and plasma membranes, suggesting a role as a high-affinity link between the actin cytoskeleton and the membrane. The encoded protein appears to aid in both myosin II assembly during cell spreading and disassembly of focal adhesions. Two transcript variants encoding different isoforms of supervillin have been described. [provided by RefSeq

Sequence: LIHAGLEPLTFTNMFPSWEHREDIAEITEMDTEVSNQITLVEDVLAKLCKTIYPLADLLARPLPEGVDPLKLEIYLTDEDFEFALDMTRDEYNALPAWKQVNLKKAKG

Specifications

Antigen SVIL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SVIL.
Formulation 50% glycerol
Gene SVIL
Gene Accession No. NM_003174
Gene Alias DKFZp686A17191
Gene Symbols SVIL
Host Species Mouse
Immunogen SVIL (NP_003165, 1679 a.a. ∽ 1786 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6840
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.