Learn More
Description
Specifically detects α-syn (aa 115-122) clone: LB509 in Human samples, and it is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
| Antigen | α-syn (aa 115-122) |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | LB509 |
| Formulation | Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Accession No. | P37840 |
| Gene Symbols | SNCA; NACP; PARK1 |
| Host Species | Mouse |
| Immunogen | Conjugated linear peptide corresponding to human α-syn (aa 115-122). |
| Purification Method | Protein G Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.