Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terminal Transferase (TdT) Mouse anti-Human, Alexa Fluor 647, Clone: E17-1519, BD

Catalog No. BDB565231 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565231 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565231 Supplier BD Biosciences Supplier No. 565231
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The E17-1519 monoclonal antibody specifically recognizes Terminal deoxynucleotidyl transferase (TdT), which is also known as Terminal transferase, or DNA nucleotidylexotransferase (DNTT). TdT is a 60 kDa nuclear polymerase that catalyzes the template-independent addition of deoxynucleotides to the 3'-hydroxyl termini of single-stranded oligonucleotide primer segments. TdT can contribute to antigen receptor diversity by N region diversification of rearranging immunoglobulin heavy chain and T cell receptor gene segments during pre-B and pre-T cell development. It is also expressed by certain lymphomas and leukemias.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Terminal Transferase (TdT)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone E17-1519
Conjugate Alexa Fluor 647
Description TdT; DNTT; Terminal transferase; Terminal addition enzyme
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Human TdT
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.