Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM189-UBE2V1 (Kua-UEV), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107786 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-786 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-786 Supplier Abnova Corporation Supplier No. H00387522A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant Kua-UEV.

The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined. [provided by RefSeq

Sequence: VHWGADTWGSVELPIVGKAFIRPFREHHIDPTAITRHDFIETNGDNCLVTLLPLLNMAYKFRTHSPEALEQ

Specifications

Antigen TMEM189-UBE2V1 (Kua-UEV)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant Kua-UEV.
Formulation 50% glycerol
Gene TMEM189-UBE2V1
Gene Accession No. NM_199203
Gene Alias CROC-1B/Kua-UEV/UBE2V1
Gene Symbols TMEM189-UBE2V1
Host Species Mouse
Immunogen Kua-UEV (NP_954673, 94 a.a. ∽ 164 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387522
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.