Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transferrin, Mouse, Clone: HTF-14, Abnova™

Catalog No. 89135525 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-525 Supplier Abnova Corporation Supplier No. MAB3878
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Transferrin.

Specifications

Antigen Transferrin
Applications ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HTF-14
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Transferrin.
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunocytochemistry Radioimmunoassay Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Porcine transferrin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes an epitope located in the N-terminal domain of humanserum transferrin, a 77 KDa single polypeptide chain glycoprotein (member of the iron binding family of proteins). It is synthesised in the liver and consists of two domains each having a high affinity reversible binding site for Fe3+.
Target Species Human, Pig, Rabbit
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.