Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF365, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106528 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-528 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-528 Supplier Abnova Corporation Supplier No. H00022891A01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse polyclonal antibody raised against a partial recombinant ZNF365.

Sequence: GLPTSDTKASFEAHVREKFNRMVEAVDRTIEKRIDKLTKELAQKTAELLEVRAAFVQLTQKKQEVQRRERALN

Specifications

Antigen ZNF365
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ZNF365.
Formulation 50% glycerol
Gene ZNF365
Gene Accession No. NM_014951
Gene Alias KIAA0844/MGC41821/MGC87345/Su48/UAN/ZNF365D
Gene Symbols ZNF365
Host Species Mouse
Immunogen ZNF365 (NP_055766, 147 a.a. ∽ 219 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22891
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.