Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse APRIL Recombinant Protein, PeproTech®

Catalog No. 315131MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-13-1MG 1 mg
31513100UG 100 μg
315-13-20UG 20 μg
31513250UG 250 μg
31513500UG 500 μg
315-13-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 315-13-1MG Supplier Gibco™ Supplier No. 315131MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine APRIL is a soluble 21.9 kDa protein, consisting of 192 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9D777
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 69583
Molecular Weight (g/mol) 21.9 kDa
Name Mouse APRIL
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 2310026N09Rik; A proliferation-inducing ligand; APRIL; CD256; TALL2; TALL-2; TNF- and APOL-related leukocyte expressed ligand 2; TNF-related death ligand 1; Tnfsf13; TNLG7B; Trdl1; TRDL-1; tumor necrosis factor (ligand) superfamily, member 13; tumor necrosis factor ligand 7 B; tumor necrosis factor ligand superfamily member 13; tumor necrosis factor superfamily member 13; tumor necrosis factor-like protein ZTNF2; tumor necrosis factor-related death ligand-1; UNQ383/PRO715; ZTNF2
Common Name APRIL
Gene Symbol TNFSF13
Biological Activity Measured by its ability to induce cell proliferation of activated T cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRREVSRLQR SGGPSQKQGE RPWQSLWEQS PDVLEAWKDG AKSRRRRAVL TQKHKKKHSV LHLVPVNITS KDSDVTEVMW QPVLRRGRGL EAQGDIVRVW DTGIYLLYSQ VLFHDVTFTM GQVVSREGQG RRETLFRCIR SMPSDPDRAY NSCYSAGVFH LHQGDIITVK IPRANAKLSL SPHGTFLGFV KL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.