Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Artemin Recombinant Protein, PeproTech®

Catalog No. 45058250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45058250UG 250 μg
45058100UG 100 μg
450-58-1MG 1 mg
450-58-20UG 20 μg
45058500UG 500 μg
450-58-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45058250UG Supplier Gibco™ Supplier No. 45058250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine Artemin is a disulfide-linked homodimer formed by two identical 113 amino acid subunits with a calculated molecular weight of 24.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z0L2
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 11876
Molecular Weight (g/mol) 12.1 kDa
Name Mouse Artemin
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor
Common Name Artemin
Gene Symbol ARTN
Biological Activity Determined by the dose-dependent stimulation of the proliferation of SH-SY5Y cells. The expected ED50 for this effect is 0.1-0.5 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AGTRSSRART TDARGCRLRS QLVPVSALGL GHSSDELIRF RFCSGSCRRA RSQHDLSLAS LLGAGALRSP PGSRPISQPC CRPTRYEAVS FMDVNSTWRT VDHLSATACG CLG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.