Learn More
Abcam Mouse AST ELISA Kit (Aspartate Aminotransferase), 1 x 96 Tests
Supplier:  Abcam AB2638821X96TESTS
Encompass_Preferred
Mouse AST ELISA kit is a single-wash, 90-minute SimpleStep ELISA assay designed for the quantification of mouse aspartate aminotransferase (AST/cAspAT) with a sensitivity of 39 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA with a 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Design your own immunoassay: we also offer the conjugation-ready, carrier-dree antibody pair. - Cited in over 25 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2015922
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.