Learn More
Biomatik Corporation Mouse ATPase, H+/K+ Exchanging Alpha Polypeptide (ATP4a) ELISA Kit
Supplier:  Biomatik Corporation EKU02630
Encompass_Preferred
Mouse ATPase, H+/K+ Exchanging Alpha Polypeptide (ATP4a) ELISA Kit for detecting ATP4a in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91WH7. Gene ID: 11944. Target Synonyms: Gastric H,K-ATPase Alpha Subunit; H (+)-K (+)-ATPase Alpha Subunit; Proton Pump; Potassium-transporting ATPase alpha chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9110
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.