Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Axl Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7121589
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8541G025 25 μg
FAB8541G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB8541G025 Supplier R&D Systems Supplier No. FAB8541G025UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Axl Monoclonal specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Axl
Applications Flow Cytometry
Classification Monoclonal
Clone 175128
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1.0 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q00993
Gene Alias AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL
Gene Symbols AXL
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Axl Ala19-Trp445 Accession # Q00993
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 558
Test Specificity Detects mouse Axl in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.