Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse BD-3 Recombinant Protein, PeproTech®

Catalog No. 2504120UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
250-41-20UG 20 μg
25041100UG 100 μg
250-41-1MG 1 mg
25041250UG 250 μg
25041500UG 500 μg
250-41-50UG 50 μg
250-41-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 250-41-20UG Supplier Gibco™ Supplier No. 2504120UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine BD-3 is a 4.6 kDa protein containing 41 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Molecular Weight (g/mol) 4.6 kDa
Name Mouse BD-3
Quantity 20 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3
Common Name beta-3 Defensin
Conjugate Unconjugated
Recombinant Recombinant
Sequence KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.