Learn More
Description
- Bladder tissue lysate was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin
- Tissue and cell debris was removed by centrifugation
- Protein concentration was determined with Bio-Rad protein assay
- The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 50 mM DTT.
Specifications
Specifications
| Quantity | 0.1 mg |
| Tissue Condition | Normal |
| Host Species | Mouse |
| Concentration | 2 mg/mL |
| Protein State | Denatured |
| Donor Source | Single Donor |
| Gender | Unavailable |
| Physical Form | Frozen |
| Tissue | Bladder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
