Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse brain cerebellum tissue lysate - total protein, 1 mg

Catalog No. 502692508
Encompass_Preferred
Catalog No. 50-269-2508 Supplier Abcam Supplier No. AB301731MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse brain cerebellum cell/tissue lysate.

  • Species: Mouse
  • Tissue: brain cerebellum
  • Form: Liquid

Specifications

For Use With (Application) Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.