Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse BRAK (CXCL14) Recombinant Protein, PeproTech®

Catalog No. 25045250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25045250UG 250 μg
25045100UG 100 μg
250-45-1MG 1 mg
250-45-20UG 20 μg
25045500UG 500 μg
250-45-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 25045250UG Supplier Gibco™ Supplier No. 25045250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine BRAK (CXCL14) is a 9.4 kDa protein containing 77 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9WUQ5
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57266
Molecular Weight (g/mol) 9.4 kDa
Name Mouse BRAK (CXCL14)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1110031L23Rik; 1200006I23Rik; AI414372; B-cell and monocyte-activating chemokine; BMAC; bolekine; BRAK; breast and kidney; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 14; chemokine BRAK; CXC; CXC chemokine in breast and kidney; CXC motif chemokine; C-X-C motif chemokine 14; C-X-C motif chemokine ligand 14; CXCL; Cxcl14; hypothetical protein LOC511771; Kec; Kidney-expressed chemokine CXC; KS1; MGC10687; MIP-2 gamma; MIP2G; MIP-2 g; MIP2gamma; musculus CXC chemokine MIP-2 gamma; NJAC; PSEC0212; SCYB14; small inducible cytokine B14; small inducible cytokine subfamily B (Cys-X-Cys), member 14; small inducible cytokine subfamily B (Cys-X-Cys), member 14 (BRAK); small-inducible cytokine B14; tumor-suppressing chemokine; UNQ240/PRO273
Common Name CXCL14
Gene Symbol Cxcl14
Biological Activity Determined by its ability to chemoattract human THP-1 monocytes supplemented with Anti-CXCL14.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIVTTKS MSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.