Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse C1qR1/CD93 Quantikine ELISA Kit
SDP

Catalog No. MCD930 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MCD930 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MCD930 Supplier R&D Systems™ Supplier No. MCD930
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 3.5 hour solid phase ELISA designed to measure mouse C1q R1 levels in cell culture supernates, cell lysates, serum, and plasma.

The Mouse C1q R1/CD93 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse C1qR1/CD93 in cell culture supernates (50 ul), cell lysates (25 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 11.4pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Mouse C1q R1
For Use With (Application) ELISA
Gene ID (Entrez) 17064
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (25uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym AA4 Antigen, C1q R1, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Target C1qR1/CD93
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.