Learn More
Abcam Mouse C1QA Synthetic Peptide, >70% Purity, 1 mg
Description
- Protein Name: C1QA
- Type: Synthetic blocking peptide
- Purity: >70%
- Species Reactivity: Mouse
- Tested Applications: BL
- Form: Lyophilized
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | C1qa (Mouse) |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Label or Dye | Unconjugated |
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Quantity | 1 mg |
| For Use With (Application) | Blocking, Neutralization |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.